Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252347.1 | NP_001239276.1 | 2 | 429 | ||
NM_016759.4 | NP_058039.1 | 2 | 429 | |||
GenBank mRNA | BC046319.1 | - | 2 | 329 | 6391042 | |
BY258909.1 | - | 2 | 402 | |||
U73941.1 | - | 2 | 156 |