Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164156.1 | NP_001157628.1 | 8 | 956 | ||
NM_016766.3 | NP_058046.2 | 9 | 1076 | |||
GenBank mRNA | AF015309.1 | - | 9 | 1026 | ||
AK088116.1 | - | 9 | 1041 | |||
AK158347.1 | - | 9 | 1056 | |||
AK158611.1 | - | 5 | 683 | |||
AK167242.1 | - | 8 | 916 | |||
AK181698.1 | - | 4 | 374 | |||
AK184558.1 | - | 4 | 374 | |||
AK209104.1 | - | 2 | 344 | |||
BC003746.1 | - | 9 | 1060 | 3486681 | ||
BC085099.1 | - | 9 | 1044 | 6391692 | ||
BC108340.1 | - | 9 | 1051 | 4504579 |