Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016778.2 | NP_058058.1 | 5 | 1108 | ||
GenBank mRNA | AF027707.1 | - | 5 | 1106 | ||
AK156243.1 | - | 5 | 1143 | |||
AK168373.1 | - | 5 | 1039 | |||
AK171258.1 | - | 5 | 1065 | |||
BC030069.1 | - | 5 | 1046 | 2936938 |