Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016782.2 | NP_058062.2 | 18, 19 | 3143 | ||
GenBank mRNA | AF039833.1 | - | 18, 19 | 3153 | ||
AK154468.1 | - | 3904 |