Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016783.3 | NP_058063.2 | 3 | 952 | ||
GenBank mRNA | AF042491.1 | - | 3 | 903 | ||
AK153272.1 | - | 3 | 969 | |||
AK159101.1 | - | 3 | 969 | |||
AK159345.1 | - | 3 | 969 | |||
AK159599.1 | - | 3 | 969 | |||
AK162315.1 | - | 3 | 972 | |||
AK166240.1 | - | 3 | 969 | |||
AK167655.1 | - | 3 | 969 | |||
AK168387.1 | - | 3 | 968 | |||
AK168882.1 | - | 3 | 969 | |||
AK169063.1 | - | 4 | 912 | |||
BC006016.1 | - | 3 | 952 | 3591536 |