Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016784.3 | NP_058064.2 | 2 | 210 | ||
GenBank mRNA | AF044334.1 | - | 1 | 102 | ||
AK075993.1 | - | 2 | 198 | |||
AK076007.1 | - | 2 | 188 | |||
AK077197.1 | - | 2 | 211 | |||
AK080068.1 | - | 2 | 217 | |||
BC006750.1 | - | 2 | 180 | 3709134 | ||
BC052895.1 | - | 2 | 169 |