Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110147.1 | NP_001103617.1 | 10, 11 | 1934 | ||
NM_001289443.1 | NP_001276372.1 | 2, 3 | 321 | |||
NM_016788.3 | NP_058068.2 | 10, 11 | 1934 | |||
GenBank mRNA | AF037260.1 | - | 10, 11 | 1717 | ||
AK087965.1 | - | 7, 8 | 1091 | |||
BC031168.1 | - | 2, 3 | 328 | 4984168 | ||
BC052421.1 | - | 10, 11 | 1934 | 6400209 | ||
DQ666696.1 | - | 10, 11 | 1745 |