Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016792.4 | NP_058072.2 | 4, 5 | 683 | ||
GenBank mRNA | AK029807.1 | - | 4, 5 | 684 | ||
AK051698.1 | - | 4, 5 | 658 | |||
AK144211.1 | - | 4, 5 | 640 | |||
AK150326.1 | - | 4, 5 | 646 | |||
AK150887.1 | - | 4, 5 | 652 | |||
AK150983.1 | - | 4, 5 | 652 | |||
AK152101.1 | - | 4, 5 | 652 | |||
AK167954.1 | - | 4, 5 | 654 | |||
AK168712.1 | - | 4, 5 | 647 | |||
AK209072.1 | - | 2, 3 | 250 | |||
AK212711.1 | - | 2, 3 | 265 | |||
BC061123.1 | - | 4, 5 | 710 | 6596299 |