Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016792.4 | NP_058072.2 | 5, 6 | 755 | ||
GenBank mRNA | AF052660.1 | - | 5, 6 | 584 | ||
AK029807.1 | - | 5, 6 | 756 | |||
AK051698.1 | - | 5, 6 | 730 | |||
AK144211.1 | - | 5, 6 | 712 | |||
AK150326.1 | - | 5, 6 | 718 | |||
AK150887.1 | - | 5, 6 | 724 | |||
AK150983.1 | - | 5, 6 | 724 | |||
AK152101.1 | - | 5, 6 | 724 | |||
AK167954.1 | - | 5, 6 | 726 | |||
AK168712.1 | - | 5, 6 | 719 | |||
BC061123.1 | - | 5, 6 | 782 | 6596299 |