Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016803.3 | NP_058083.2 | 4 | 3445 | ||
GenBank mRNA | BC055055.1 | - | 2 | 3324 | 6493234 |