Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016804.4 | NP_058084.3 | 8 | 662 | ||
GenBank mRNA | AB041562.1 | - | 8 | 654 | ||
AF053550.1 | - | 8 | 635 | |||
AK005233.1 | - | 8 | 594 | |||
AK083055.1 | - | 624 | ||||
AK087933.1 | - | 8 | 650 | |||
AK168000.1 | - | 8 | 655 | |||
AK168021.1 | - | 8 | 625 | |||
AK168045.1 | - | 8 | 654 | |||
AK169048.1 | - | 8 | 658 | |||
AK208117.1 | - | 5 | 283 | |||
AK209045.1 | - | 4 | 294 | |||
AK211256.1 | - | 5 | 320 | |||
AK214537.1 | - | 5 | 283 | |||
AK214538.1 | - | 5 | 283 | |||
AK218920.1 | - | 5 | 283 | |||
AK220102.1 | - | 5 | 283 | |||
BC006641.1 | - | 8 | 612 | 3483318 |