Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252600.1 | NP_001239529.1 | 5 | 854 | ||
NM_001252601.1 | NP_001239530.1 | 5 | 854 | |||
NM_016850.3 | NP_058546.1 | 5 | 854 | |||
GenBank mRNA | AK079685.1 | - | 5 | 854 | ||
AK089869.1 | - | 5 | 854 | |||
AK150403.1 | - | 5 | 854 | |||
AK150608.1 | - | 5 | 854 | |||
AK151470.1 | - | 5 | 854 | |||
AK152401.1 | - | 5 | 854 | |||
AK152403.1 | - | 5 | 854 | |||
AK153054.1 | - | 5 | 854 | |||
AK155950.1 | - | 1 | 1658 | |||
AK158378.1 | - | 5 | 853 | |||
AK158403.1 | - | 5 | 853 | |||
AK159883.1 | - | 5 | 854 | |||
BC138799.1 | - | 4 | 794 | 8861821 | ||
DT923384.1 | - | 3 | 331 | |||
U73037.1 | - | 4 | 831 |