Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016856.3 | NP_058552.1 | 3 | 433 | ||
GenBank mRNA | AF012822.1 | - | 3 | 419 | ||
AK054372.1 | - | 3 | 416 | |||
AK140869.1 | - | 3 | 415 | |||
AK141325.1 | - | 3 | 415 | |||
AK147746.1 | - | 3 | 434 | |||
BC007163.1 | - | 1 | 91 | |||
BC013628.1 | - | 3 | 408 | 3497854 |