Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016856.3 | NP_058552.1 | 12, 13 | 2023 | ||
GenBank mRNA | AF012822.1 | - | 12, 13 | 2009 | ||
BC007163.1 | - | 10, 11 | 1681 | |||
BC013628.1 | - | 12, 13 | 1998 | 3497854 |