Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016859.3 | NP_058555.3 | 7 | 1833 | ||
GenBank mRNA | AK013468.1 | - | 3 | 744 | ||
AK019239.1 | - | 7 | 1568 | |||
AK032993.1 | - | 7 | 1565 | |||
AK049195.1 | - | 7 | 1826 | |||
AK051704.1 | - | 7 | 1823 | |||
AK083033.1 | - | 7 | 1820 | |||
AK143027.1 | - | 7 | 1834 | |||
AK168301.1 | - | 7 | 1571 | |||
BC017530.1 | - | 7 | 1560 | 4925307 |