Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016861.4 | NP_058557.2 | 4 | 546 | ||
GenBank mRNA | AF053367.2 | - | 4 | 462 | ||
AK076285.1 | - | 445 | ||||
AK088089.1 | - | 4 | 445 | |||
AK146225.1 | - | 4 | 456 | |||
AK146399.1 | - | 4 | 458 | |||
AK158173.1 | - | 4 | 445 | |||
AK189667.1 | - | 2 | 77 | |||
BC004809.1 | - | 4 | 437 | 3588132 |