Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vti1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293685.1 | NP_001280614.1 | 9 | 2174 | ||
NM_001293686.1 | NP_001280615.1 | 8 | 2164 | |||
NM_016862.4 | NP_058558.1 | 8 | 2153 | |||
GenBank mRNA | AK004751.1 | - | 8 | 1849 | ||
AK028646.1 | - | 8 | 1859 | |||
AK148281.1 | - | 8 | 1893 | |||
BC012211.1 | - | 8 | 1851 | |||
BC089321.1 | - | 9 | 1881 | 6839437 |