Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282072.1 | NP_001269001.1 | 2 | 526 | ||
NM_001282073.1 | NP_001269002.1 | 3 | 450 | |||
NM_001282076.1 | NP_001269005.1 | 3 | 483 | |||
NM_016874.3 | NP_058570.1 | 2 | 526 | |||
GenBank mRNA | AF102818.1 | - | 2 | 386 | ||
AK082115.1 | - | 2 | 379 | |||
AK144333.1 | - | 2 | 295 | |||
AK146546.1 | - | 2 | 403 | |||
AK155836.1 | - | 2 | 492 | |||
AK158586.1 | - | 3 | 335 | |||
BC046399.1 | - | 3 | 428 | 5063995 | ||
BY051636.1 | - | 2 | 209 | |||
FJ377318.1 | - | 3 | 343 | |||
FJ377319.1 | - | 2 | 371 |