Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281929.1 | NP_001268858.1 | 1, 2 | 352 | ||
NM_001290451.1 | NP_001277380.1 | 1, 2 | 124 | |||
NM_016886.4 | NP_058582.3 | 1, 2 | 352 | |||
GenBank mRNA | AB022342.1 | - | 1, 2 | 142 | ||
AB079072.1 | - | 1, 2 | 88 | |||
AB102775.1 | - | 1, 2 | 88 | |||
AB102776.1 | - | 1, 2 | 88 | |||
AF483494.1 | - | 1, 2 | 88 | |||
AF483495.1 | - | 1, 2 | 88 | |||
AK135043.1 | - | 1, 2 | 105 | |||
AK138615.1 | - | 1, 2 | 257 | |||
AK157815.1 | - | 2, 3 | 529 | |||
AK161163.1 | - | 1, 2 | 172 | |||
AK220530.1 | - | 1, 2 | 139 | |||
BC076584.1 | - | 1, 2 | 217 | |||
BC086678.1 | - | 1, 2 | 297 | 30668476 | ||
BC129855.1 | - | 1, 2 | 142 | 40109751 | ||
BC129856.1 | - | 1, 2 | 142 | 40109752 | ||
BY266507.1 | - | 1, 2 | 354 | |||
CJ107988.1 | - | 1, 2 | 350 |