Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281929.1 | NP_001268858.1 | 8 | 1383 | ||
NM_001290451.1 | NP_001277380.1 | 8 | 1155 | |||
NM_016886.4 | NP_058582.3 | 8 | 1383 | |||
GenBank mRNA | AB022342.1 | - | 8 | 1173 | ||
AB079072.1 | - | 8 | 1119 | |||
AB102775.1 | - | 8 | 1119 | |||
AB102776.1 | - | 8 | 1119 | |||
AF483494.1 | - | 8 | 1119 | |||
AF483495.1 | - | 8 | 1119 | |||
AK135043.1 | - | 8 | 1136 | |||
AK138615.1 | - | 8 | 1288 | |||
AK157815.1 | - | 9 | 1560 | |||
AK161163.1 | - | 8 | 1203 | |||
AK220530.1 | - | 8 | 1170 | |||
BC076584.1 | - | 8 | 1248 | |||
BC086678.1 | - | 8 | 1328 | 30668476 | ||
BC129855.1 | - | 8 | 1173 | 40109751 | ||
BC129856.1 | - | 8 | 1173 | 40109752 |