Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016906.4 | NP_058602.1 | 12 | 2357 | ||
GenBank mRNA | AK007696.1 | - | 1 | 719 | ||
AK077849.1 | - | 1 | 866 | |||
AK088473.1 | - | 12 | 2358 | |||
AK153055.1 | - | 12 | 2347 | |||
AK159349.1 | - | 12 | 2343 | |||
AK167484.1 | - | 12 | 2286 | |||
BC003707.1 | - | 12 | 2283 | 3592772 | ||
BC006020.1 | - | 8 | 1942 |