Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199023.1 | NP_001185952.1 | 17 | 3391 | ||
NM_001199024.1 | NP_001185953.1 | 16 | 3226 | |||
NM_001199025.1 | NP_001185954.1 | 17 | 2731 | |||
NM_016915.4 | NP_058611.1 | 16 | 2772 | |||
GenBank mRNA | AF259401.1 | - | 16 | 2671 | ||
AK144495.1 | - | 17 | 2605 | |||
AK153333.1 | - | 16 | 3032 | |||
AK193793.1 | - | 1 | 104 | |||
BC003487.1 | - | 16 | 2565 | 2811357 | ||
BC049778.1 | - | 16 | 2952 | |||
BC052845.1 | - | 16 | 2597 | 30054573 | ||
BC057209.1 | - | 17 | 2984 | 5329147 |