Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016963.2 | NP_058659.1 | 7 | 880 | ||
GenBank mRNA | AF177172.1 | - | 7 | 868 | ||
AF237630.1 | - | 7 | 794 | |||
AK017725.1 | - | 7 | 876 | |||
AK050372.1 | - | 7 | 882 | |||
AK084463.1 | - | 4 | 402 | |||
AK147017.1 | - | 7 | 881 | |||
AK167723.1 | - | 7 | 878 | |||
AK169218.1 | - | 7 | 881 | |||
AK210770.1 | - | 2 | 127 | |||
BC082595.1 | - | 7 | 872 | 30645500 |