Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290745.1 | NP_001277674.1 | 6 | 705 | ||
NM_016965.3 | NP_058661.1 | 5 | 687 | |||
GenBank mRNA | AJ534525.1 | - | 5 | 470 | ||
AK122318.1 | - | 5 | 557 | |||
AK197674.1 | - | 1 | 26 | |||
AK219064.1 | - | 1 | 35 | |||
BC060197.1 | - | 5 | 666 | |||
BC127067.1 | - | 6 | 487 |