Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093764.1 | NP_001087233.1 | 3 | 910 | ||
NM_001093765.1 | NP_001087234.1 | 3 | 859 | |||
NM_001093766.1 | NP_001087235.1 | 2 | 797 | |||
NM_016969.2 | NP_058665.2 | 3 | 698 | |||
GenBank mRNA | AJ001616.1 | - | 3 | 719 | ||
AK052258.1 | - | 3 | 817 | |||
AK053414.1 | - | 3 | 650 | |||
AK089538.1 | - | 3 | 681 | |||
AK149962.1 | - | 3 | 841 | |||
AK159160.1 | - | 841 | ||||
AK180990.1 | - | 1 | 145 | |||
AK197378.1 | - | 1 | 157 | |||
BC056355.1 | - | 3 | 793 | 6811773 | ||
BC119364.1 | - | 2 | 581 | 8734113 | ||
BC119368.1 | - | 2 | 578 | 8734117 |