Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016985.2 | NP_058681.1 | 3 | 482 | ||
GenBank mRNA | AF073997.1 | - | 3 | 455 | ||
AY099894.1 | - | 3 | 180 | |||
AY099895.1 | - | 4 | 207 | |||
AY099896.1 | - | 5 | 258 | |||
AY099897.1 | - | 3 | 207 | |||
AY099898.1 | - | 4 | 231 | |||
BC056376.1 | - | 3 | 482 | 6849757 | ||
BC057337.1 | - | 3 | 476 | 6825916 |