Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017378.2 | NP_059074.2 | 1 | 627 | ||
GenBank mRNA | AK048100.1 | - | 1 | 626 | ||
Y08715.1 | - | 1 | 592 |