Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017401.2 | NP_059097.2 | 11 | 1792 | ||
GenBank mRNA | AF176098.1 | - | 11 | 1565 | ||
AJ251804.2 | - | 11 | 1536 | |||
AK189181.1 | - | 2 | 164 | |||
BC056213.1 | - | 11 | 1584 | 30024844 | ||
BC056996.1 | - | 8 | 3439 |