Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113517.1 | NP_001106989.1 | 7 | 941 | ||
NM_001113518.1 | NP_001106990.1 | 6 | 730 | |||
NM_017402.4 | NP_059098.2 | 6 | 730 | |||
GenBank mRNA | AF247654.1 | - | 6 | 626 | ||
AF247655.1 | - | 4 | 347 | |||
AF343877.1 | - | 6 | 610 | |||
AK052545.1 | - | 6 | 701 | |||
AK129064.1 | - | 7 | 877 | |||
AK139156.1 | - | 7 | 870 | |||
AY220301.1 | - | 6 | 695 | |||
BC044838.1 | - | 6 | 715 | 3499604 | ||
U96634.1 | - | 4 | 420 |