Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017407.2 | NP_059103.1 | 2 | 161 | ||
GenBank mRNA | AF420307.1 | - | 2 | 98 | ||
AK052228.1 | - | 2 | 129 | |||
AK076619.1 | - | 2 | 157 | |||
BC052672.1 | - | 2 | 125 | 30058496 | ||
X81633.1 | - | 2 | 97 |