Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017470.2 | NP_059498.2 | 4 | 835 | ||
GenBank mRNA | AB010031.1 | - | 4 | 813 | ||
AK002657.1 | - | 4 | 785 | |||
AK132061.1 | - | 4 | 825 | |||
AK207401.1 | - | 1 | 139 | |||
BC005426.1 | - | 4 | 766 | 3593932 |