Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 4 | 788 | ||
GenBank mRNA | AB017616.1 | - | 4 | 711 | ||
AK150792.1 | - | 4 | 742 | |||
AK159355.1 | - | 4 | 721 | |||
AK159434.1 | - | 4 | 743 | |||
AK166658.1 | - | 4 | 734 | |||
BC005417.1 | - | 4 | 736 | 3495022 | ||
BC037732.1 | - | 1 | 85 | |||
BC071245.1 | - | 4 | 1136 |