Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168568.1 | NP_001162040.1 | 19 | 2418 | ||
NM_001168569.1 | NP_001162041.1 | 19 | 2475 | |||
NM_001168571.1 | NP_001162043.1 | 17 | 2397 | |||
NM_018737.5 | NP_061207.1 | 19 | 2550 | |||
GenBank mRNA | AK032018.1 | - | 19 | 2277 | ||
AK052854.1 | - | 19 | 2466 | |||
AK084595.1 | - | 19 | 2347 | |||
AK085381.1 | - | 19 | 2326 | |||
AK164206.1 | - | 17 | 2224 |