Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253700.1 | NP_001240629.1 | 4, 5 | 435 | ||
NM_018740.2 | NP_061210.2 | 6, 7 | 917 | |||
GenBank mRNA | AF004107.1 | - | 5, 6 | 836 | ||
AK011130.1 | - | 4, 5 | 729 | |||
AK013662.1 | - | 4, 5 | 518 | |||
AK152582.1 | - | 6, 7 | 921 | |||
BC003752.1 | - | 5, 6 | 768 | 3489878 | ||
BE862527.1 | - | 1, 2 | 193 |