Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102458.1 | NP_001095928.1 | 5 | 947 | ||
NM_001301688.1 | NP_001288617.1 | 4 | 787 | |||
NM_018745.5 | NP_061215.1 | 4 | 787 | |||
GenBank mRNA | AF032128.1 | - | 2 | 213 | ||
AK049680.1 | - | 4 | 788 | |||
AK088112.1 | - | 4 | 785 | |||
AK088671.1 | - | 5 | 944 | |||
AK133914.1 | - | 4 | 783 | |||
BC019412.1 | - | 4 | 775 | 5151406 | ||
BC021152.1 | - | 4 | 740 | |||
BC043722.1 | - | 2 | 293 | 5321733 | ||
BC046814.1 | - | 4 | 690 |