Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006757.3 | NP_006748.1 | 5, 7 | 289 | ||
XM_006718290.3 | XP_006718353.1 | 7, 9 | 228 | |||
XM_011520343.2 | XP_011518645.1 | 7, 9 | 359 | |||
XM_017018206.1 | XP_016873695.1 | 7, 9 | 151 | |||
XM_017018207.1 | XP_016873696.1 | 6, 8 | 139 | |||
XM_017018208.1 | XP_016873697.1 | 5, 7 | 106 | |||
XM_017018216.1 | XP_016873705.1 | 6, 8 | 210 | |||
XM_017018217.1 | XP_016873706.1 | 5, 7 | 189 | |||
XM_017018218.1 | XP_016873707.1 | 4, 6 | 177 | |||
GenBank mRNA | M21984.1 | - | 4, 6 | 89 | ||
U14641.1 | - | 118 | ||||
U14642.1 | - | 85 |