Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018749.2 | NP_061219.2 | 8 | 823 | ||
GenBank mRNA | AB012580.1 | - | 8 | 764 | ||
AK045228.1 | - | 8 | 765 | |||
AK049767.1 | - | 8 | 762 | |||
AK145689.1 | - | 8 | 778 | |||
AK168256.1 | - | 8 | 813 | |||
AK183958.1 | - | 61 | ||||
AK185859.1 | - | 1 | 20 | |||
BC018598.1 | - | 8 | 761 | |||
BC089020.1 | - | 8 | 757 | 30551991 |