Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018750.3 | NP_061220.2 | 2, 3 | 617 | ||
GenBank mRNA | AF053959.1 | - | 2, 3 | 599 | ||
AK029321.1 | - | 2, 3 | 619 | |||
AK155534.1 | - | 2, 3 | 613 | |||
BC089605.1 | - | 2, 3 | 619 | 30121804 |