Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018753.6 | NP_061223.2 | 6 | 1087 | ||
GenBank mRNA | AK002632.1 | - | 6 | 1106 | ||
AK004872.1 | - | 6 | 1088 | |||
AK011389.1 | - | 6 | 1087 | |||
AK083367.1 | - | 6 | 1077 | |||
AK144061.1 | - | 6 | 1150 | |||
AK150414.1 | - | 6 | 1100 | |||
AK151294.1 | - | 6 | 1100 | |||
AK158932.1 | - | 6 | 990 | |||
BC049070.1 | - | 921 | ||||
BC132121.1 | - | 6 | 941 | 40130398 | ||
BC138512.1 | - | 6 | 941 | 8861532 |