Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018759.2 | NP_061229.2 | 5 | 340 | ||
GenBank mRNA | AB012725.1 | - | 5 | 306 | ||
AK017693.1 | - | 5 | 341 | |||
AK146438.1 | - | 5 | 321 | |||
AV503563.1 | - | 5 | 351 | |||
BC005567.1 | - | 5 | 331 | |||
BC039041.1 | - | 4 | 2345 |