Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018759.2 | NP_061229.2 | 5 | 523 | ||
GenBank mRNA | AB012725.1 | - | 5 | 489 | ||
AK017693.1 | - | 5 | 524 | |||
AK146438.1 | - | 5 | 504 | |||
BC005567.1 | - | 5 | 514 | |||
BC039041.1 | - | 4 | 2528 |