Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018759.2 | NP_061229.2 | 6, 7 | 926 | ||
GenBank mRNA | AB012725.1 | - | 6, 7 | 892 | ||
AK017693.1 | - | 6, 7 | 927 | |||
AK146438.1 | - | 6, 7 | 907 | |||
BC005567.1 | - | 6, 7 | 917 | |||
BC037055.1 | - | 4, 5 | 467 | 4948371 | ||
BC039041.1 | - | 5, 6 | 2931 | |||
BC042769.1 | - | 1, 2 | 1822 | |||
BU503985.1 | - | 3, 4 | 310 |