Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018763.2 | NP_061233.2 | 2 | 2569 | ||
GenBank mRNA | AB011451.1 | - | 2 | 2021 | ||
AB011452.1 | - | 2 | 2021 | |||
AK131820.1 | - | 1 | 649 | |||
BC051963.1 | - | 2 | 1956 | 5707543 |