Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195130.1 | NP_001182059.1 | 8 | 1504 | ||
NM_018774.4 | NP_061244.1 | 8 | 1497 | |||
GenBank mRNA | AF060076.1 | - | 1 | 83 | ||
AK083804.1 | - | 3 | 1644 | |||
BC057571.1 | - | 8 | 1440 | 6410302 | ||
U81491.1 | - | 7 | 1394 |