Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195083.1 | NP_001182012.1 | 7 | 2339 | ||
NM_001195130.1 | NP_001182059.1 | 15 | 3739 | |||
NM_018774.4 | NP_061244.1 | 15 | 3732 | |||
GenBank mRNA | AB062362.1 | - | 7 | 2325 | ||
BC051657.1 | - | 7 | 2205 | |||
BC057571.1 | - | 15 | 3675 | 6410302 | ||
BC071246.1 | - | 7 | 2319 | |||
U81491.1 | - | 14 | 3629 |