Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018777.4 | NP_061247.1 | 3 | 942 | ||
GenBank mRNA | AK010560.1 | - | 3 | 943 | ||
AK010682.1 | - | 3 | 937 | |||
BC005718.1 | - | 3 | 919 | 3594823 | ||
BC050138.1 | - | 3 | 925 | 30028814 |