Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018788.3 | NP_061258.2 | 3 | 903 | ||
GenBank mRNA | AK220344.1 | - | 1 | 246 | ||
BC058526.1 | - | 1 | 293 | |||
BC065073.1 | - | 1 | 368 | 6834580 |