Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018788.3 | NP_061258.2 | 7 | 5917 | ||
GenBank mRNA | AK019308.1 | - | 1 | 201 | ||
AK137842.1 | - | 5 | 4996 | |||
BC038525.1 | - | 1 | 622 | |||
BC058526.1 | - | 5 | 5308 | |||
BC065073.1 | - | 5 | 5382 | 6834580 |