Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hils1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018792.1 | NP_061262.1 | 1 | 333 | ||
GenBank mRNA | AB022320.1 | - | 1 | 333 | ||
AB045724.1 | - | 1 | 326 | |||
AK005718.1 | - | 1 | 364 | |||
AY286319.1 | - | 1 | 333 | |||
BC050966.1 | - | 1 | 366 | |||
BC061119.1 | - | 1 | 396 | 6773493 |