Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018815.2 | NP_061285.2 | 3 | 431 | ||
GenBank mRNA | AF113751.1 | - | 3 | 399 | ||
AK053417.1 | - | 3 | 432 | |||
AK157272.1 | - | 3 | 423 | |||
AK173056.1 | - | 3 | 385 | |||
AK202156.1 | - | 123 | ||||
BC052468.1 | - | 3 | 424 | 6835038 |